Stampa…

DL-Homophenylalanine ≥97.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: 2-Amino-4-phenylbutyric Acid, H-DL-HomoPhe-OH, H-DL-HoPhe-OH

H1329-1G H1329-5G
TCIAH1329-1GEA 52 EUR
TCIAH1329-1G TCIAH1329-5G
DL-Homophenylalanine ≥97.0% (by HPLC, titration analysis)
DL-Homophenylalanine
Formula: C₁₀H₁₃NO₂
Peso molecolare: 179,22 g/mol
Punto di fusione: 294°C
MDL Number: MFCD00063092
CAS Number: 1012-05-1

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Nonaqueous Titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR