4-(2-Hydroxyethyl)cyclohexanol (cis- and trans- mixture) cis- and trans-mixture ≥98.0% (by GC)
Fornitore: TCI
Formula:
C₈H₁₆O₂ Punto di ebollizione: 175°C |
CAS Number:
74058-21-2 |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % (total of isomers) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...