Stampa…

trans-4-(Hydroxymethyl)cyclohexanecarboxylic acid 98,0 GC, T

Fornitore: TCI
Attenzione

H1243-1G H1243-5G
TCIAH1243-1GEA 90 EUR
TCIAH1243-1G TCIAH1243-5G
trans-4-(Hydroxymethyl)cyclohexanecarboxylic acid 98,0 GC, T
trans-4-(Hydroxymethyl)cyclohexanecarboxylic acid
Formula: C₈H₁₄O₃
Peso molecolare: 158.20 g/mol
MDL Number: MFCD03840884
CAS Number: 66185-74-8

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 139.0 to 143.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR