cis-4-(Hydroxymethyl)cyclohexanecarboxylic acid ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
H1242-25G
H1242-5G
H1242-1G
TCIAH1242-25GEA
559
EUR
TCIAH1242-25G
TCIAH1242-5G
TCIAH1242-1G
cis-4-(Hydroxymethyl)cyclohexanecarboxylic acid ≥98.0% (by GC)
cis-4-(Hydroxymethyl)cyclohexanecarboxylic acid
Formula:
C₈H₁₄O₃ Peso molecolare: 158,20 g/mol |
CAS Number:
73094-35-6 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 97.0 % |
Melting point | 117.0 to 121.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...