1,2,3,4,8,9,10,11,15,16,17,18,22,23,24,25-Hexadecafluorophthalocyaninecopper(II) ≥98.0% (by total nitrogen and titration analysis), purified by sublimation
Fornitore: TCI
Synonyms:
F16CuPc, Copper(II) 1,2,3,4,8,9,10,11,15,16,17,18,22,23,24,25-Hexadecafluorophthalocyanine
Formula:
C₃₂CuF₁₆N₈ Peso molecolare: 863.93 g/mol |
MDL Number:
MFCD00274642 CAS Number: 14916-87-1 |
Risultati del test di specificazione
Appearance | Orange to Brown to Dark purple powder to crystal |
Purity (with Total Nitrogen) | min. 98.0 % |
Purity (Chelometric Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...