Stampa…

Hexadecyl acrylate ≥90.0% (by GC) stabilizzato

Fornitore: TCI EUROPE
Attenzione

Synonyms: Acrylic Acid Hexadecyl Ester

Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

H1168-25G H1168-250G
TCIAH1168-25GEA 42 EUR
TCIAH1168-25G TCIAH1168-250G
Hexadecyl acrylate ≥90.0% (by GC) stabilizzato
Hexadecyl acrylate
Formula: C₁₉H₃₆O₂
Peso molecolare: 296,50 g/mol
Flash Pt: 178°C
MDL Number: MFCD00078441
CAS Number: 13402-02-3
EINECS: 236-492-0

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to lump
Purity (GC) min. 90.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR