2-[4-(Hydroxymethyl)-2-nitrophenoxy]ethanol ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
Ethylene Glycol Mono-4-(hydroxymethyl)-2-nitrophenyl Ether, 4-(2-Hydroxyethoxy)-3-nitrobenzyl Alcohol
H1048-1G
H1048-100MG
TCIAH1048-1GEA
374
EUR
TCIAH1048-1G
TCIAH1048-100MG
2-[4-(Hydroxymethyl)-2-nitrophenoxy]ethanol ≥98.0% (by GC)
2-[4-(Hydroxymethyl)-2-nitrophenoxy]ethanol
Risultati del test di specificazione
Appearance | Light yellow to Yellow to Green powder to crystal |
Solubility in Methanol | almost transparency |
Purity (GC) | min. 98.0 % |
Melting point | 48.0 to 52.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...