Stampa…

1-(2-Hydroxyethyl)-3,3-dimethylindolino-6'-nitrobenzopyrylospiran ≥93.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: 1'-(2-Hydroxyethyl)-3',3'-dimethyl-6-nitrospiro[1(2H)-benzopyran-2,2'-indoline], 2-(3',3'-Dimethyl-6-nitrospiro[chromene-2,2'-indolin]-1'-yl)ethanol

H1042-1G H1042-5G
TCIAH1042-1GEA 98 EUR
TCIAH1042-1G TCIAH1042-5G
1-(2-Hydroxyethyl)-3,3-dimethylindolino-6'-nitrobenzopyrylospiran ≥93.0% (by HPLC, titration analysis)
1-(2-Hydroxyethyl)-3,3-dimethylindolino-6'-nitrobenzopyrylospiran
Formula: C₂₀H₂₀N₂O₄
Peso molecolare: 352,39 g/mol
MDL Number: MFCD00369031
CAS Number: 16111-07-2

Visualizza i codici

Risultati del test di specificazione

Appearance Orange to Brown to Dark purple powder to crystal
Purity (HPLC) min. 93.0 area%
Purity (Nonaqueous Titration) min. 93.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR