Stampa…
N-Hydroxy-4-nitrophthalimide ≥98.0% (by HPLC)
Fornitore: TCI EUROPE
Attenzione
Synonyms: 4-Nitrophthaloxime
H1036-5G H1036-1G
TCIAH1036-5GEA 196 EUR
TCIAH1036-5G TCIAH1036-1G
N-Hydroxy-4-nitrophthalimide ≥98.0% (by HPLC)
N-Hydroxy-4-nitrophthalimide
Formula: C₈H₄N₂O₅
Peso molecolare: 208,13 g/mol
MDL Number: MFCD01115771
CAS Number: 105969-98-0
Visualizza i codici
Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 95.0 %
Melting point 167.0 to 171.0 deg-C