Stampa…

Ethyl (R)-(-)-3-hydroxybutyrate ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: Ethyl (R)-3-hydroxybutyrate, (R)-(-)-3-Hydroxybutyric Acid Ethyl Ester

H1029-1G H1029-5G
TCIAH1029-1GEA 32 EUR
TCIAH1029-1G TCIAH1029-5G
Ethyl (R)-(-)-3-hydroxybutyrate ≥98.0% (by GC)
Ethyl (R)-(-)-3-hydroxybutyrate
Formula: C₆H₁₂O₃
Peso molecolare: 132,16 g/mol
Flash Pt: 64°C
MDL Number: MFCD00075386
CAS Number: 24915-95-5

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Specific rotation [a]20/D -16.0 to -20.0 deg (neat)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR