Stampa…

N-(Hydroxymethyl)nicotinamide ≥97.0% (by HPLC, titration analysis)

Fornitore: TCI
Attenzione

Synonyms: N-(Hydroxymethyl)-3-pyridinecarboxamide

H1001-25G H1001-500G
TCIAH1001-25GEA 27 EUR
TCIAH1001-25G TCIAH1001-500G
N-(Hydroxymethyl)nicotinamide ≥97.0% (by HPLC, titration analysis)
N-(Hydroxymethyl)nicotinamide
Formula: C₇H₈N₂O₂
MDL Number: MFCD00010437
CAS Number: 3569-99-1
EINECS: 222-668-4
Merck Index: 14,04833

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 143.0 to 147.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR