Stampa…

2-(3-Hydroxypropyl)benzimidazole ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE

Synonyms: 3-(2-Benzimidazolyl)propanol, 3-(1H-Benzo[d]imidazol-2-yl)propan-1-ol, 2-Benzimidazolepropanol

H0988-25G H0988-5G
TCIAH0988-25GEA 184 EUR
TCIAH0988-25G TCIAH0988-5G
2-(3-Hydroxypropyl)benzimidazole ≥98.0% (by titrimetric analysis)
2-(3-Hydroxypropyl)benzimidazole
Formula: C₁₀H₁₂N₂O
Peso molecolare: 176,22 g/mol
MDL Number: MFCD00022688
CAS Number: 2403-66-9
EINECS: 219-290-7

Visualizza i codici

Risultati del test di specificazione

Appearance White to Brown powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 165.0 to 169.0 deg-C
Solubility in Methanol very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR