(S)-2-Amino-4-phenylbutanoic acid ≥98.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
Synonyms:
H-HoPhe-OH, H-Homophe-OH, (S)-2-Amino-4-phenylbutyric Acid
H0985-1G
H0985-100MG
TCIAH0985-1GEA
34
EUR
TCIAH0985-1G
TCIAH0985-100MG
(S)-2-Amino-4-phenylbutanoic acid ≥98.0% (by titrimetric analysis)
(S)-2-Amino-4-phenylbutanoic acid
Formula:
C₁₀H₁₃NO₂ Peso molecolare: 179,22 g/mol |
MDL Number:
MFCD00002619 CAS Number: 943-73-7 EINECS: 213-403-3 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Specific rotation [a]20/D | +43.0 to +47.0 deg (C=1, 3mol/L HCl) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...