Stampa…

2-Hexyl-4-pentynoic acid ≥95.0% (by titrimetric analysis)

Fornitore: TCI
Attenzione

H0964-25G H0964-5G
TCIAH0964-25GEA 651 EUR
TCIAH0964-25G TCIAH0964-5G
2-Hexyl-4-pentynoic acid ≥95.0% (by titrimetric analysis)
2-Hexyl-4-pentynoic acid
Formula: C₁₁H₁₈O₂
MDL Number: MFCD00946863
CAS Number: 96017-59-3

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (Neutralization titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR