5-Hydroxy-2-adamantanone ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
Idramantone, 5-Hydroxyadamantan-2-one
H0962-1G
H0962-25G
H0962-5G
TCIAH0962-1GEA
36
EUR
TCIAH0962-1G
TCIAH0962-25G
TCIAH0962-5G
5-Hydroxy-2-adamantanone ≥98.0% (by GC)
5-Hydroxy-2-adamantanone
Formula:
C₁₀H₁₄O₂ Peso molecolare: 166,22 g/mol |
MDL Number:
MFCD00192211 CAS Number: 20098-14-0 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...