Stampa…

(R)-(+)-ɑ-Hydroxy-γ-butyrolactone ≥98.0% (by GC)

Fornitore: TCI

H0950-1G H0950-100MG
TCIAH0950-1GEA 1444 EUR
TCIAH0950-1G TCIAH0950-100MG
(R)-(+)-ɑ-Hydroxy-γ-butyrolactone ≥98.0% (by GC)
(R)-(+)-alpha-Hydroxy-gamma-butyrolactone
Formula: C₄H₆O₃
Punto di ebollizione: 133°C
MDL Number: MFCD00211246
CAS Number: 56881-90-4

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Optical purity (GC) min. 96.0 ee%
Specific rotation [a]20/D +67.0 to +76.0 deg (C=1, CHCl3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR