3-Hydroxyaspartic acid ≥98.0% (by total nitrogen and titration analysis)
Fornitore: TCI
Formula:
C₄H₇NO₅ |
MDL Number:
MFCD00128126 CAS Number: 71653-06-0 EINECS: 275-771-1 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (with Total Nitrogen) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...