Stampa…

Hexyl methyl ether ≥98.0% (by GC)

Fornitore: TCI
Pericolo

Synonyms: N-hexylmethylether

H0930-25ML H0930-100ML
TCIAH0930-25MLEA 25 EUR
TCIAH0930-25ML TCIAH0930-100ML
Hexyl methyl ether ≥98.0% (by GC)
Hexyl methyl ether
Formula: C₇H₁₆O
Punto di ebollizione: 125°C
Flash Pt: 20°C
MDL Number: MFCD00015350
CAS Number: 4747-07-3
EINECS: 225-263-0
UN: 3271
ADR: 3,II

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR