Stampa…

4-Hydroxy-4-phenylpiperidine ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 4-Phenyl-4-piperidinol

H0925-25G H0925-5G
TCIAH0925-25GEA 267 EUR
TCIAH0925-25G TCIAH0925-5G
4-Hydroxy-4-phenylpiperidine ≥98.0% (by GC, titration analysis)
4-Hydroxy-4-phenylpiperidine
Formula: C₁₁H₁₅NO
Peso molecolare: 177,25 g/mol
MDL Number: MFCD00006000
CAS Number: 40807-61-2
EINECS: 255-089-0

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 157.0 to 162.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR