Haloperidol ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
4-[4-(4-Chlorophenyl)-4-hydroxypiperidino]-4'-fluorobutyrophenone
H0912-25G
H0912-5G
TCIAH0912-25GEA
244
EUR
TCIAH0912-25G
TCIAH0912-5G
Haloperidol ≥98.0% (by HPLC, titration analysis)
Haloperidol
Formula:
C₂₁H₂₃ClFNO₂ Peso molecolare: 375,87 g/mol |
MDL Number:
MFCD00051423 CAS Number: 52-86-8 EINECS: 200-155-6 UN: 2811 ADR: 6.1,III Merck Index: 14,04598 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Solubility in Acetic acid | almost transparency |
Melting point | 150.0 to 153.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...