Methyl-3-hydroxy-2,2-dimethylpropanoate ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Hydroxypivalic acid methyll ester, Methyl 2,2-Dimethyl-3-hydroxypropionate, Methyl 3-hydroxy-2,2-dimethylpropanoate, Methyl Hydroxypivalate, Methyl 2,2-dimethyl-3-hydroxypropionate, 2,2-Dimethyl-3-hydroxypropionic acid methyll ester
H0906-25G
H0906-500G
TCIAH0906-25GEA
47
EUR
TCIAH0906-25G
TCIAH0906-500G
Methyl-3-hydroxy-2,2-dimethylpropanoate ≥98.0% (by GC)
Methyl-3-hydroxy-2,2-dimethylpropanoate
Formula:
C₆H₁₂O₃ Peso molecolare: 132,1600 g/mol Punto di ebollizione: 178°C Flash Pt: 77°C |
MDL Number:
MFCD00009707 CAS Number: 14002-80-3 EINECS: 237-805-3 |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...