Ethyl-2-hydroxy-4-(trimethylsilyl)-3-butynoate ≥97.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
2-Hydroxy-4-(trimethylsilyl)-3-butynoic Acid Ethyl Ester
H0895-1G
H0895-5G
TCIAH0895-1GEA
100
EUR
TCIAH0895-1G
TCIAH0895-5G
Ethyl-2-hydroxy-4-(trimethylsilyl)-3-butynoate ≥97.0% (by GC)
Ethyl-2-hydroxy-4-(trimethylsilyl)-3-butynoate
Formula:
C₉H₁₆O₃Si Peso molecolare: 200,31 g/mol Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00671544 CAS Number: 66697-09-4 |
Risultati del test di specificazione
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...