Stampa…

Hexadecafluorosebacic acid ≥96.0% (by GC, titration analysis)

Fornitore: TCI
Attenzione

H0892-25G H0892-5G
TCIAH0892-25GEA 407 EUR
TCIAH0892-25G TCIAH0892-5G
Hexadecafluorosebacic acid ≥96.0% (by GC, titration analysis)
Hexadecafluorosebacic acid
Formula: C₁₀H₂F₁₆O₄
MDL Number: MFCD00042365
CAS Number: 307-78-8

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 96.0 %
Purity (Neutralization titration) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR