Stampa…

(S)-(+)-5-Hydroxymethyl-2-pyrrolidinone ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

H0867-1G H0867-5G
TCIAH0867-1GEA 19 EUR
TCIAH0867-1G TCIAH0867-5G
(S)-(+)-5-Hydroxymethyl-2-pyrrolidinone ≥98.0% (by GC)
(S)-(+)-5-Hydroxymethyl-2-pyrrolidinone
Formula: C₅H₉NO₂
Peso molecolare: 115,13 g/mol
MDL Number: MFCD00077792
CAS Number: 17342-08-4

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (with Total Nitrogen) min. 97.0 %
Melting point 82.0 to 87.0 deg-C
Specific rotation [a]20/D +30.0 to +32.0 deg (C=5, EtOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR