Hydrazine carbonate ∼70% ∼7.3 mol/L in water
Fornitore: TCI
|
Attenzione
|
H0854-25G
H0854-500G
TCIAH0854-25GEA
16
EUR
TCIAH0854-25G
TCIAH0854-500G
Hydrazine carbonate ∼70% ∼7.3 mol/L in water
Hydrazine carbonate
Formula:
CH₈N₄O₂ |
MDL Number:
MFCD00185569 CAS Number: 112077-84-6 |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Concentration (Potassium iodate Method) | 70.0 to 77.0 w/w% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...