Stampa…

meso-Hydrobenzoin ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: meso-1,2-Diphenylethylene glycol, meso-1,2-Diphenyl-1,2-ethanediol, rel-(1R,2S)-1,2-Diphenylethane-1,2-diol

H0816-25G H0816-5G
TCIAH0816-25GEA 148 EUR
TCIAH0816-25G TCIAH0816-5G
meso-Hydrobenzoin ≥98.0% (by GC)
meso-Hydrobenzoin
Formula: C₁₄H₁₄O₂
Peso molecolare: 214,26 g/mol
MDL Number: MFCD00064253
CAS Number: 579-43-1
Merck Index: 14,04777

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 133.0 to 137.0 deg-C
Solubility in Methanol very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR