Stampa…

5-(4-Hydroxyphenyl)hydantoin ≥98.0% (by total nitrogen basis)

Fornitore: TCI EUROPE

Synonyms: 5-(4-Hydroxyphenyl)imidazolidine-2,4-dione

H0806-25G H0806-5G
TCIAH0806-25GEA 175 EUR
TCIAH0806-25G TCIAH0806-5G
5-(4-Hydroxyphenyl)hydantoin ≥98.0% (by total nitrogen basis)
5-(4-Hydroxyphenyl)hydantoin
Formula: C₉H₈N₂O₃
Peso molecolare: 192,17 g/mol
MDL Number: MFCD00044002
CAS Number: 2420-17-9
EINECS: 219-340-8

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR