2-(Hydroxymethyl)anthracene ≥98.0% (by HPLC)
Fornitore: TCI EUROPE
Synonyms:
2-Anthracenemethanol
H0750-1G
H0750-5G
TCIAH0750-1GEA
229
EUR
TCIAH0750-1G
TCIAH0750-5G
2-(Hydroxymethyl)anthracene ≥98.0% (by HPLC)
2-(Hydroxymethyl)anthracene
Formula:
C₁₅H₁₂O Peso molecolare: 208,26 g/mol |
MDL Number:
MFCD00142932 CAS Number: 22863-82-7 |
Risultati del test di specificazione
Appearance | Light yellow to Yellow to Orange powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Melting point | 221.0 to 227.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...