Stampa…

2-(5-tert-Butyl-2-hydroxyphenyl)benzotriazole ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

H0719-25G H0719-500G
TCIAH0719-25GEA 34 EUR
TCIAH0719-25G TCIAH0719-500G
2-(5-tert-Butyl-2-hydroxyphenyl)benzotriazole ≥98.0% (by titrimetric analysis)
2-(5-tert-Butyl-2-hydroxyphenyl)benzotriazole
Formula: C₁₆H₁₇N₃O
Peso molecolare: 267,33 g/mol
MDL Number: MFCD03844795
CAS Number: 3147-76-0
EINECS: 221-574-0

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 96.0 to 100.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR