Stampa…

4-Hydroxy-6-methyl-2-pyrone 98,0 GC_ASSAY_METHOD

Fornitore: TCI
Attenzione

Synonyms: 4-Hydroxy-6-methyl-2H-pyran-2-one, 3,5-Dihydroxysorbic acid d-lactone, 2-hydroxy-6-methyl-pyran-4-one

H0715-25G H0715-500G
TCIAH0715-25GEA 41 EUR
TCIAH0715-25G TCIAH0715-500G
4-Hydroxy-6-methyl-2-pyrone 98,0 GC_ASSAY_METHOD
4-Hydroxy-6-methyl-2-pyrone
Formula: C₆H₆O₃
Peso molecolare: 126.11 g/mol
MDL Number: MFCD00006641
CAS Number: 675-10-5
EINECS: 211-619-2

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR