Stampa…

α-n-Hexylcinnamaldehyde ≥90.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2-Hexyl-3-phenyl-2-propenal, α-Hexylcinnamaldehyde, alpha-Hexylcinnamaldehyde, 2-Hexyl-3-phenylacrylaldehyde

H0685-25ML H0685-500ML
TCIAH0685-25MLEA 17 EUR
TCIAH0685-25ML TCIAH0685-500ML
α-n-Hexylcinnamaldehyde ≥90.0% (by GC)
α-n-Hexylcinnamaldehyde
Formula: C₁₅H₂₀O
Peso molecolare: 216,3187 g/mol
Punto di ebollizione: 175°C
MDL Number: MFCD00006989
CAS Number: 101-86-0
EINECS: 202-983-3

Visualizza i codici

Risultati del test di specificazione

Appearance White or Colorless to Yellow powder to lump to clear liquid
Purity (GC) min. 90.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR