3-Indolepropanol ≥98.0% (by GC)
Fornitore: TCI
Formula:
C₁₁H₁₃NO Punto di ebollizione: 200°C |
MDL Number:
MFCD00013437 CAS Number: 3569-21-9 EINECS: 222-667-9 |
Risultati del test di specificazione
Appearance | White to Yellow to Green clear liquid to slightly cloudy liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...