Homogentisic acid-γ-lactone 98,0 GC_ASSAY_METHOD
Fornitore: TCI
|
Attenzione
|
Synonyms:
2,3-Dihydro-5-hydroxybenzofuran-2-one, 2,5-Dihydroxyphenylacetic Acid γ-Lactone, 5-Hydroxybenzofuran-2(3H)-one, 5-Hydroxy-2-coumaranone
Formula:
C₈H₆O₃ Peso molecolare: 150.13 g/mol |
MDL Number:
MFCD00005857 CAS Number: 2688-48-4 EINECS: 220-252-7 |
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 191.0 to 194.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...