Stampa…

2-Hydroxy-n-octanoic acid ≥98.0% (by titrimetric analysis)

Fornitore: TCI
Attenzione

Synonyms: 2-Hydroxy-n-octanoic Acid, 2-Hydroxyoctanoic acid

H0592-25G H0592-5G
TCIAH0592-25GEA 270 EUR
TCIAH0592-25G TCIAH0592-5G
2-Hydroxy-n-octanoic acid ≥98.0% (by titrimetric analysis)
2-Hydroxy-n-octanoic acid
Formula: C₈H₁₆O₃
Peso molecolare: 160.21 g/mol
MDL Number: MFCD00014410
CAS Number: 617-73-2
EINECS: 210-524-3

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 95.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 68.0 to 71.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR