2-(5-Chloro-2-benzotriazolyl)-6-tert-butyl-p-cresol ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
2-(tert-Butyl)-6-(5-chloro-2H-benzo[d][1,2,3]triazol-2-yl)-4-methylphenol, 2-(2-Hydroxy-3-tert-butyl-5-methylphenyl)-5-chlorobenzotriazole
H0559-25G
H0559-500G
TCIAH0559-25GEA
17
EUR
TCIAH0559-25G
TCIAH0559-500G
2-(5-Chloro-2-benzotriazolyl)-6-tert-butyl-p-cresol ≥98.0% (by GC)
2-(5-Chloro-2-benzotriazolyl)-6-tert-butyl-p-cresol
Formula:
C₁₇H₁₈ClN₃O Peso molecolare: 315,80 g/mol |
MDL Number:
MFCD00059707 CAS Number: 3896-11-5 EINECS: 223-445-4 |
Risultati del test di specificazione
Appearance | Light yellow to Yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 140.0 to 143.0 deg-C |
Solubility in Toluene | within almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...