Nickel(II) hexafluoroacetylacetonate hydrate
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
Hexafluoroacetylacetono Nickel(II) Salt, Bis(hexafluoroacetylacetonato)nickel(II) hydrate
H0558-1G
H0558-5G
TCIAH0558-1GEA
49
EUR
TCIAH0558-1G
TCIAH0558-5G
Nickel(II) hexafluoroacetylacetonate hydrate
Nickel(II) hexafluoroacetylacetonate hydrate
Formula:
C₁₀H₂F₁₂NiO₄·xH₂O Peso molecolare: 472,80 g/mol (anhy) |
MDL Number:
MFCD00067506 CAS Number: 14949-69-0 EINECS: 239-028-5 |
Risultati del test di specificazione
Appearance | Light yellow to Amber to Dark green powder to crystal |
Purity (Chelometric Titration) | min. 96.0 % (calcd.on anh.substance) |
Water | 5.0 to 13.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...