4-Hydroxy-3-methoxyphenylpyruvic acid ≥95.0% (by HPLC)
Fornitore: TCI
|
Attenzione
|
Synonyms:
Vanilpyruvic acid, 3-(4-Hydroxy-3-methoxyphenyl)-2-oxopropanoic acid
Formula:
C₁₀H₁₀O₅ Peso molecolare: 210.19 g/mol |
MDL Number:
MFCD00056707 CAS Number: 1081-71-6 |
Risultati del test di specificazione
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (HPLC) | min. 95.0 area% |
Purity (Neutralization titration) | min. 90.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...