Stampa…

Hecogenin acetate ≥90.0% (by GC)

Fornitore: TCI

H0537-1G H0537-5G
TCIAH0537-1GEA 64 EUR
TCIAH0537-1G TCIAH0537-5G
Hecogenin acetate ≥90.0% (by GC)
Hecogenin acetate
Formula: C₂₉H₄₄O₅
MDL Number: MFCD00005088
CAS Number: 915-35-5
EINECS: 213-021-7

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 90.0 %
Specific rotation [a]20/D -3.0 to -5.0 deg (C=1, Dioxane)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR