Stampa…

Hyodeoxycholic acid ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE

H0535-1G H0535-25G
TCIAH0535-1GEA 16 EUR
TCIAH0535-1G TCIAH0535-25G
Hyodeoxycholic acid ≥98.0% (by titrimetric analysis)
Hyodeoxycholic acid
Formula: C₂₄H₄₀O₄
Peso molecolare: 392,58 g/mol
MDL Number: MFCD00003681
CAS Number: 83-49-8
EINECS: 201-483-2
Merck Index: 14,04857

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 95.0 %
Purity (Neutralization titration) min. 98.0 %
Specific rotation [a]20/D +7.5 to +9.5 deg (C=4, EtOH)
Solubility in Methanol very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR