Stampa…

trans-3-Hexenedioic acid ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: trans-beta-Hydromuconic Acid, trans-2-Butene-1,4-dicarboxylic Acid, Dihydromuconic Acid, Hex-3-enedioic acid

H0477-25G H0477-5G
TCIAH0477-25GEA 193 EUR
TCIAH0477-25G TCIAH0477-5G
trans-3-Hexenedioic acid ≥98.0% (by titrimetric analysis)
trans-3-Hexenedioic acid
Formula: C₆H₈O₄
Peso molecolare: 144,13 g/mol
MDL Number: MFCD00002785
CAS Number: 4436-74-2
EINECS: 224-648-0

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 196.0 to 199.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR