1-Hydroxyethylferrocene ≥95.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
1-Ferrocenylethanol, (1-Hydroxyethyl)ferrocene, α-Hydroxyethylferrocene
H0463-1G
H0463-5G
TCIAH0463-1GEA
27
EUR
TCIAH0463-1G
TCIAH0463-5G
1-Hydroxyethylferrocene ≥95.0% (by titrimetric analysis)
1-Hydroxyethylferrocene
Formula:
C₁₂H₅FeO Peso molecolare: 221,01 g/mol |
MDL Number:
MFCD00013752 CAS Number: 1277-49-2 |
Risultati del test di specificazione
Appearance | Light yellow to Brown powder to crystal |
Purity (Chelometric Titration) | min. 95.0 % |
Melting point | 76.0 to 79.0 deg-C |
Solubility in Toluene | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...