1-Hydrazinophthalazine hydrochloride ≥99.0% (by HPLC, titration analysis)
Fornitore: TCI
|
Pericolo
|
Synonyms:
Hydralazine Hydrochloride, Hydralazine HCl, 1-Hydrazinylphthalazine hydrochloride
Formula:
C₈H₈N₄ Peso molecolare: 196.64 g/mol Punto di fusione: 273°C |
MDL Number:
MFCD00135998 CAS Number: 304-20-1 EINECS: 206-151-0 UN: 2811 ADR: 6.1 III Merck Index: 14,04763 |
|
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 99.0 area% |
Purity (Potassium iodate Method) | min. 99.0 % |
Solubility in Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...