Hydroxydicyclopentadiene (so called) ≥90.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
8(9)-Hydroxytricyclo[5.2.1.0(2,6)]dec-3-ene
H0384-25ML
H0384-500ML
TCIAH0384-25MLEA
16
EUR
TCIAH0384-25ML
TCIAH0384-500ML
Hydroxydicyclopentadiene (so called) ≥90.0% (by GC)
Hydroxydicyclopentadiene
Formula:
C₁₀H₁₄O Peso molecolare: 150,22 g/mol Flash Pt: 106°C |
MDL Number:
MFCD00151152 CAS Number: 37275-49-3 EINECS: 253-436-0 |
Risultati del test di specificazione
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 90.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...