Stampa…

Het anhydride ≥95.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Chlorendic Anhydride, 1,4,5,6,7,7-Hexachloro-5-norbornene-2,3-dicarboxylic anhydride, 1,4,5,6,7,7-Hexachloro-5-norbornene-2,3-dicarboxylic Anhydride

H0349-25G H0349-500G
TCIAH0349-25GEA 31 EUR
TCIAH0349-25G TCIAH0349-500G
Het anhydride ≥95.0% (by titrimetric analysis)
Het anhydride
Formula: C₉H₂Cl₆O₃
Peso molecolare: 370,81 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00080438
CAS Number: 115-27-5
EINECS: 204-077-3
Merck Index: 14,02086

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 95.0 %
Melting point 235.0 to 240.0 deg-C
Solubility in Toluene very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR