Stampa…

3-Hydroxy-N-(1-naphthyl)-2-naphthamide

Fornitore: TCI EUROPE

Synonyms: Azoic Coupling Component 4, Naphthol AS-BO, 3-Hydroxy-N-naphthalen-1-ylnaphthalene-2-carboxamide, 2-Hydroxy-3-naphthoic Acid 1-Naphthylamide

H0318-25G H0318-500G
TCIAH0318-25GEA 16 EUR
TCIAH0318-25G TCIAH0318-500G
3-Hydroxy-N-(1-naphthyl)-2-naphthamide
3-Hydroxy-N-(1-naphthyl)-2-naphthamide
Formula: C₂₁H₁₅NO₂
Peso molecolare: 313,36 g/mol
MDL Number: MFCD00043900
CAS Number: 132-68-3
EINECS: 205-075-5

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light red powder to crystal

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR