7-Hydroxy-5-methyl[1,2,4]triazolo[1,5-a]pyrimidine
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
7-Hydroxy-5-methyl-1,3,4-triazaindolizine, 5-Methyl-[1,2,4]triazolo[1,5-a]pyrimidin-7-ol, 4-Hydroxy-6-methyl-1,3,3a,7-tetraazaindene
H0273-25G
H0273-500G
TCIAH0273-25GEA
45
EUR
TCIAH0273-25G
TCIAH0273-500G
7-Hydroxy-5-methyl[1,2,4]triazolo[1,5-a]pyrimidine
7-Hydroxy-5-methyl[1,2,4]triazolo[1,5-a]pyrimidine
Formula:
C₆H₆N₄O Peso molecolare: 150,14 g/mol Punto di fusione: 226°C |
MDL Number:
MFCD00005555 CAS Number: 2503-56-2 EINECS: 219-706-7 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | min. 98.0 % |
Solubility in hot Water | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...