Methyl-2-hydroxyisobutyrate ≥99.5% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Methyl 2-Hydroxyisobutyrate, 2-Hydroxyisobutyric Acid Methyl Ester
H0256-25ML
H0256-500ML
H0256-100ML
TCIAH0256-25MLEA
19
EUR
TCIAH0256-25ML
TCIAH0256-500ML
TCIAH0256-100ML
Methyl-2-hydroxyisobutyrate ≥99.5% (by GC)
Methyl-2-hydroxyisobutyrate
Formula:
C₅H₁₀O₃ Peso molecolare: 118,13 g/mol Punto di ebollizione: 137°C Flash Pt: 45°C |
MDL Number:
MFCD00004457 CAS Number: 2110-78-3 EINECS: 218-301-2 UN: 1993 ADR: 3,III |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 99.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...