2-Hydrazinoquinoline ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
2-Quinolylhydrazine, 2-Hydrazinylquinoline
H0178-1G
H0178-25G
H0178-5G
TCIAH0178-1GEA
50
EUR
TCIAH0178-1G
TCIAH0178-25G
TCIAH0178-5G
2-Hydrazinoquinoline ≥98.0% (by HPLC, titration analysis)
2-Hydrazinoquinoline
Formula:
C₉H₉N₃ Peso molecolare: 159,19 g/mol |
MDL Number:
MFCD00041860 CAS Number: 15793-77-8 EINECS: 239-899-1 |
Risultati del test di specificazione
Appearance | Light yellow to Brown powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 140.0 to 145.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...