DL-Homocysteic acid ≥97.0% (by titrimetric analysis)
Fornitore: TCI
Formula:
C₄H₉NO₅S |
MDL Number:
MFCD00007536 CAS Number: 504-33-6 EINECS: 207-991-0 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...