Stampa…

2,5-Hexanediol ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Hexane-2,5-diol, 2,5-Dihydroxyhexane

H0100-25ML H0100-5ML
TCIAH0100-25MLEA 95 EUR
TCIAH0100-25ML TCIAH0100-5ML
2,5-Hexanediol ≥98.0% (by GC)
2,5-Hexanediol
Formula: C₆H₁₄O₂
Peso molecolare: 118,18 g/mol
Flash Pt: 110°C
MDL Number: MFCD00004557
CAS Number: 2935-44-6
EINECS: 220-910-3

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR